Traceable® Infrared Thermometers with Laser and Calibration
Small, lightweight thermometers take instant temperature readings
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • Laser-sighting pinpoints your measurement area for accurate measurements
  • Backlighting permits viewing in low light conditions
  • °F/°C selectable
Simple one-handed point-and-shoot operation makes it easy to use anywhere you need to take temperature measurements. Noninvasive measurements are ideal for food preparation, life sciences, and pharmaceutical applications.
Traceable® Products have an individually-numbered Traceable® Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.
15,900.00 - ₹18,100.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Distance To Target Size Ratio (D:S) Emissivity Temperature Input
Availability Pricing
COMPARE
11:1 Fixed at 0.95 None
₹15,900.00INR / Each
COMPARE
12:1 Adjustable from 0.10 to 1.00 Thermocouple type K probe
₹18,100.00INR / Each