The Determination of the Metastable Zone Width - Case Study Lysozyme
Introduction
 |
| Figure 1. Cole-Parmer Integrity 10 Reaction Station |
Phase diagrams are fundamental to the design and development of optimized crystallization processes, ensuring the production of high-quality crystals. The measurement of a sample solutions solubility and nucleation point can take a long time when using classical techniques. In the case of proteins, this is a particular problem as the determination cannot be carried out simultaneously by one single method.
In this application note, a rapid, reliable, and reproducible determination method to determine the metastable zone width of lysozyme in sodium acetate buffer solution is presented. The method uses a Cole-Parmer Integrity 10 Reaction Station to allow transmission detectors the saturation and crystallization points to be determined using the turbidity measurement technique.
Experimental Methods
Hen egg white lysozyme was purchased from Fluka (CAS-No 9001-63-2) and dissolved in 0.1 M sodium acetate buffer pH 5.0 and 4% (wt) sodium chloride (NaCl) to give solutions with concentrations ranging from 10 mg/ml to 40 mg/ml.
The solutions were heated and cooled in the Cole-Parmer Integrity 10 Reaction Station in a controlled manner at 0.1°C/min to determine the nucleation and solubility points. Turbidity measurements were collected using the optional in-stu IR probes as shown on the right.
Results
The solubility point was defined as the point at which the % transmission reached a stable plateau, and the nucleation point was defined as the first point at which a sustained drop in % transmission was measured, as shown in Figure 2.
Figure 2. Turbidity changes of lysozyme solution with temperature (15 mg/ml).
The nucleation and solubility points were determined for several solution concentrations as shown in Figure 3.
Figure 3. Turbidity changes of lysozyme solution with temperature; 12 mg/ml and 20 mg/ml.
The nucleation and solubility points, as determined from the turbidity measurements, were plotted along with literature data for the solubility of lysozyme in the pseudo-phase diagram shown in Figure 4.
Figure 4. Pseudo-phase diagram of lysozyme protein (4% NaCl, 0.1 M Sodium acetate buffer pH 5.0).
The construction of a diagram of this type allowed the metastable zone to be easily identified.
Conclusion
The nucleation and solubility points for systems with different protein concentrations were determined using a turbidity measurement technique. The method is characterized by good reproducibility and reliability. The results from this investigation were coupled with data reported in literature to extend the known phase diagram and the data obtained in this study showed a good correlation. The use of this technique allows the metastable zone width to be determined within a few hours and with higher precision when compared to other classical methods.
Acknowledgements
The contents of this application note are taken directly from a poster presentation of the same name given by S. Maosoongnern1, D. Pertig2, V. Diaz Borbon2, J. Ulrich2.
1School of Chemical Engineering, Suranaree University of Techology, Nakhon Ratchasima 30000, Thailand
2Martin-Luther-Universitität Halle-Wittenberg, Zentrum für Ingenieurwissenschaften, Verfahrenstechnik / TVT, D-06099 Halle (Saale), Germany