Cannon® Zeitfuchs® Cross-Arm Calibrated Glass Capillary Viscometers
Determine viscosities using ASTM testing methods
  • Each viscometer is calibrated in accordance with ASTM D446
  • Comes with a calibration certificate that includes the viscometer constant and formulas for calculating viscosities
Use to determine kinematic viscosities of transparent and opaque Newtonian liquids. Require a 1- to 3-mL minimum sample volume..
81,600.00 - ₹86,800.00INR / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
Compareitem specifications
Approximate Constant (cSt/sec) Max Viscosity Range (cSt) Universal Size Number
Availability Pricing
Cannon 9723-S50 Cross-arm viscometer, size 1 WW-98934-00 Mfr # 9723-S50
COMPARE
0.003 3 1
₹81,600.00INR / Each
Cannon 9723-S53 Zeitfuchs cross-arm viscometer, size 2 WW-98934-01 Mfr # 9723-S53
COMPARE
0.01 10 2
₹86,800.00INR / Each
Cannon 9723-S56 Cross-arm viscometer, size 3 WW-98934-02 Mfr # 9723-S56
COMPARE
0.03 30 3
₹86,800.00INR / Each
Cannon 9723-S59 Cross-arm viscometer, size 4 WW-98934-03 Mfr # 9723-S59
COMPARE
0.1 100 4
₹81,600.00INR / Each
Cannon 9723-S62 Cross-arm viscometer, size 5 WW-98934-04 Mfr # 9723-S62
COMPARE
0.3 300 5
₹81,600.00INR / Each
Cannon 9723-S65 Cross-arm viscometer, size 6 WW-98934-05 Mfr # 9723-S65
COMPARE
1 1000 6
₹81,600.00INR / Each
Cannon 9723-S68 Cross-arm viscometer, size 7 WW-98934-06 Mfr # 9723-S68
COMPARE
3 3000 7
₹81,600.00INR / Each
Cannon 9723-S71 Cross-arm viscometer, size 8 WW-98934-07 Mfr # 9723-S71
COMPARE
10 10000 8
₹81,600.00INR / Each
Cannon 9723-S74 Cross-arm viscometer, size 9 WW-98934-08 Mfr # 9723-S74
COMPARE
30 30000 9
₹81,600.00INR / Each
Cannon 9723-S77 Cross-arm viscometer, size 10 WW-98934-09 Mfr # 9723-S77
COMPARE
100 100000 10
₹81,600.00INR / Each
Showing 1- 10 of 10