Cannon 9723-S53 Zeitfuchs cross-arm viscometer, size 2
Representative image only.
Cannon 9723-S53 Zeitfuchs cross-arm viscometer, size 2
Cannon | Mfr: 9723-S53| Item: 9893401
Determine viscosities using ASTM testing methods
₹108,000.00INR / Each
₹108,000.00INR / Each
Buy It Together
Current Item
Cannon 9723-S53 Zeitfuchs cross-arm viscometer, size 2
Cannon 9723-S53 Zeitfuchs cross-arm viscometer, size 2
9893401
₹108,000.00INR / Each
+ =
Total Price 117900.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Each viscometer is calibrated in accordance with ASTM D446
  • Comes with a calibration certificate that includes the viscometer constant and formulas for calculating viscosities
  • 1 year warranty
More About this Item
Use to determine kinematic viscosities of transparent and opaque Newtonian liquids. Require a 1- to 3-mL minimum sample volume..
1 Year Warranty
Specifications
  • Accuracy±0.3%
  • Viscometer TypeZeitfuchs Cross-Arm
  • Approximate Constant (cSt/sec)0.01
  • Sample Volume Needed (mL)1
  • Universal Size Number2
  • Min Viscosity Range (cSt)2
  • Max Viscosity Range (cSt)10
  • DescriptionZeitfuchs cross-arm viscometer, size 2