Cannon 9723-S68 Cross-arm viscometer, size 7
Representative image only.
Cannon 9723-S68 Cross-arm viscometer, size 7
Cannon | Mfr: 9723-S68| Item: 9893406
Determine viscosities using ASTM testing methods
₹101,000.00INR / Each
₹101,000.00INR / Each
Buy It Together
Current Item
Cannon 9723-S68 Cross-arm viscometer, size 7
Cannon 9723-S68 Cross-arm viscometer, size 7
9893406
₹101,000.00INR / Each
+ =
Total Price 110900.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Each viscometer is calibrated in accordance with ASTM D446
  • Comes with a calibration certificate that includes the viscometer constant and formulas for calculating viscosities
  • 1 year warranty
More About this Item
Use to determine kinematic viscosities of transparent and opaque Newtonian liquids. Require a 1- to 3-mL minimum sample volume..
1 Year Warranty
Specifications
  • Accuracy±0.3%
  • Viscometer TypeZeitfuchs Cross-Arm
  • Approximate Constant (cSt/sec)3
  • Sample Volume Needed (mL)1
  • Universal Size Number7
  • Min Viscosity Range (cSt)600
  • Max Viscosity Range (cSt)3000
  • DescriptionCross-arm viscometer, size 7