Zefon PTFE Filter Membranes with Support
Excellent for air monitoring and sampling in aggressive environments
  • Supports provide strength
  • PTFE membranes offer interference-free results
PTFE membranes with supports provide the chemical resistance and strength needed for sampling in harsh environments.
25,600.00 - ₹121,000.00INR / Pkg of 100
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Diameter (mm) Membrane Support Pore Size (micron)
Availability Pricing
COMPARE
25 PTFE 1
₹38,500.00INR / Pkg of 100
COMPARE
37 PTFE 1
₹25,600.00INR / Pkg of 50
COMPARE
47 PTFE 1
₹38,500.00INR / Pkg of 50
COMPARE
37 Polypropylene 1
₹106,000.00INR / Pkg of 100
COMPARE
37 Polypropylene 0.45
₹121,000.00INR / Pkg of 100
COMPARE
37 Polymethylpentene ring 2
₹113,000.00INR / Pkg of 50
COMPARE
47 Polymethylpentene ring 2
₹119,000.00INR / Pkg of 50
COMPARE
25 Polymethylpentene ring 3
₹95,500.00INR / Pkg of 50
Showing 1- 8 of 8