Zefon 37-mm Cassette Housings
Insert a filter membrane of your choice to suit your application
  • Leak-free design contains all the samples collected
  • Use with 37-mm diameter membranes
Select your housing from five materials of construction. Clear styrene provides excellent visibility, allowing you to confirm that the filter media is installed and working properly. Select opaque styrene for light-sensitive samples. Natural polypropylene (PP) offers excellent chemical resistance. Use conductive black PP housing when wall deposit is a concern; the housing becomes conductive when grounded properly. Styrene-acrylonitrile (SAN) resin is similar to styrene but with added durability.
Use two-piece housings for closed face sampling applications. With 35% less surface area than three-piece housings, there is less surface area for the contaminants to cling to, so more contaminants will end up on the filter membrane. Three-piece housings feature one spacer ring while the four-piece housings feature two spacer rings, separating the inlet from the filter media. Four-piece model allows for a center filter placement to be placed in the shoot-through FTIR cassette for on-site analysis. Both three- and four-piece housings are suitable for open and closed face sampling applications.
6,200.00 - ₹15,300.00INR / Box of 50
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
Compareitem specifications
Color Material Description
Availability Pricing
COMPARE
Clear Styrene Cassette Housing, 37 mm, 2 Piece, Clear Styrene; 50/BX
₹6,200.00INR / Box of 50
COMPARE
Black Polypropylene Cassette Housing, 37 mm, 2 Piece, Conductive Black PP; 50/BX
₹9,300.00INR / Box of 50
COMPARE
Brown Styrene Cassette Housing, 37 mm, 2 Piece, Opaque Styrene; 50/BX
₹9,300.00INR / Box of 50
COMPARE
Natural Polypropylene Cassette Housing, 37 mm, 2 Piece, Natural Polypropylene; 50/BX
₹9,300.00INR / Box of 50
COMPARE
Clear Styrene Cassette Housing, 37 mm, 3 Piece, Clear Styrene; 50/BX
₹6,600.00INR / Box of 50
COMPARE
Black Polypropylene Cassette Housing, 37 mm, 3 Piece, Conductive Black PP; 50/BX
₹9,500.00INR / Box of 50
COMPARE
Brown Styrene Cassette Housing, 37 mm, 3 Piece, Opaque Styrene; 50/BX
₹9,300.00INR / Box of 50
COMPARE
Natural Polypropylene Cassette Housing, 37 mm, 3 Piece, Natural Polypropylene; 50/BX
₹14,800.00INR / Box of 50
COMPARE
Clear Styrene-acrylonitrile (SAN) Cassette Housing, 37 mm, 4 Piece, SAN; 50/BX
₹15,300.00INR / Box of 50
Showing 1- 9 of 9