YSI EcoSense ODO200 Portable Optical DO Meter
Accurate, economical optical dissolved oxygen meter
  • No electrode maintenance — no membranes and solutions required
  • Automatic temperature compensation from 0 to 50 C
  • Manual salinity and pressure compensation for measurements
  • Saves up to 50 data sets in memory
The ODO200 meter uses optical measurement technology to provide fast response times with minimal maintenance. Add temperature compensation (ATC), barometric pressure compensation, and salinity correction to further ensure measurement accuracy. Designed for quick, accurate optical dissolved oxygen measurements in an economical platform.
Rugged IP67-rated waterproof housing makes the meter ideal for plant or field testing. Features low-batttery indicator, auto shutoff, and 50 data set memory.
321,000.00 - ₹366,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
YSI ODO200 EcoSense Optical DO Meter Kit wirh 1M cable 5935320 Mfr # 606324
COMPARE
EcoSense Optical DO Meter Kit wirh 1M cable
₹321,000.00INR / Each
YSI 606325 EcoSense ODO200 Optical DO Meter Kit w 4M cable 5935322 Mfr # 606325
COMPARE
EcoSense ODO200 Optical DO Meter Kit w 4M cable
₹339,000.00INR / Each
YSI 606305 EcoSense ODO200 Optical DO Kit w 10M cable 5935324 Mfr # 606305
COMPARE
EcoSense ODO200 Optical DO Kit w 10M cable
₹366,000.00INR / Each