Wrapping Paper for Steam & ETO
Sterilization wraps or wrapping papers keep the contents sterile packed after the sterilization process during storage prior to use. The use of two wraps reinforces the strength of the packaging and offers a further guarantee of sterility. The wraps meet the requirements of the standard sterilization procedures - steam, ethylene oxide or formaldehyde. Due to the use of cellulose fibers in the composition of its sheets, they reduce the risk of residual humidity in the packs after steam sterilization. Since the nonwoven are resistant to disinfectants, they can be also be used in sterile fields (alcohol repellent grade).
12,000.00INR / Pkg of 1
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (in) Width (in)
Availability Pricing
COMPARE
20
Call For Price
COMPARE
30
Call For Price
COMPARE
35
Call For Price
COMPARE
39
₹12,000.00INR / Pkg of 1
COMPARE
47
Call For Price
COMPARE
20
Call For Price
COMPARE
30
Call For Price
COMPARE
35
Call For Price
COMPARE
39
Call For Price
COMPARE
47
Call For Price
Showing 1- 10 of 10