Crepe Paper - Green Size Of 75 Cm X 75 Cm, 250 Papers
Representative image only.
Crepe Paper - Green Size Of 75 Cm X 75 Cm, 250 Papers
| Item: 9470234
Call For Price
Call For Price
Product Description
More About this Item
Sterilization wraps or wrapping papers keep the contents sterile packed after the sterilization process during storage prior to use. The use of two wraps reinforces the strength of the packaging and offers a further guarantee of sterility. The wraps meet the requirements of the standard sterilization procedures - steam, ethylene oxide or formaldehyde. Due to the use of cellulose fibers in the composition of its sheets, they reduce the risk of residual humidity in the packs after steam sterilization. Since the nonwoven are resistant to disinfectants, they can be also be used in sterile fields (alcohol repellent grade).
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.
Specifications
  • Width (in)30
  • Depth (in)30
  • DescriptionCrepe Paper - Green Size Of 75 Cm X 75 Cm, 250 Papers