Wildco Surber Stream Bottom Sampler
Quantitative sampler for benthic invertebrates
  • Recommended for sampling shallow streams
  • Open base encloses area of 12 x 12"
Also known as a riffle or run sampler this sampler has two pivoting stainless steel frames with lockable braces to hold the frames at right angles to each other. The net is 29" long with a filter area that is 17.5" long
103,000.00 - ₹107,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Description Material
Availability Pricing
Wildco 12-C22 Surber Stream Bottom Sampler, 243 um net 0549110 Mfr # 12-C22
COMPARE
Surber Stream Bottom Sampler, 243 um net Stainless Steel, Nitex Nylon
₹107,000.00INR / Each
Wildco 12-C32 Surber Stream Bottom Sampler, 363 um net 0549112 Mfr # 12-C32
COMPARE
Surber Stream Bottom Sampler, 363 um net Stainless Steel, Nitex Nylon
₹107,000.00INR / Each
Wildco 12-C62 Surber Stream Bottom Sampler, 600 um net 0549116 Mfr # 12-C62
COMPARE
Surber Stream Bottom Sampler, 600 um net Stainless Steel, Nitex Nylon
₹103,000.00INR / Each