Wildco 12-C22 Surber Stream Bottom Sampler, 243 um net
Representative image only.
Wildco 12-C22 Surber Stream Bottom Sampler, 243 um net
Wildco | Mfr: 12-C22| Item: 0549110
Quantitative sampler for benthic invertebrates
₹107,000.00INR / Each
₹107,000.00INR / Each
Product Description
Key Features
  • Recommended for sampling shallow streams
  • Open base encloses area of 12 x 12"
  • 3 month warranty
More About this Item
Also known as a riffle or run sampler this sampler has two pivoting stainless steel frames with lockable braces to hold the frames at right angles to each other. The net is 29" long with a filter area that is 17.5" long
3 Month Warranty
Specifications
  • MaterialStainless Steel, Nitex Nylon
  • DescriptionSurber Stream Bottom Sampler, 243 um net