Wildco Hess Stream Bottom Sampler
Quantitative sampler for benthic invertebrates
  • Recommended for sampling shallow streams (<18") with gravel, cobble bottoms
  • Open base encloses area of 0.086 square meters
This 304 SS cylindrical sampler has open top and reversible handles for pushing and rotating into stream bottoms. Once in place, scrub the substrate inside so that specimens are carried by the current into the net. Includes Dolphin™ adapter and bucket for easy sample removal. Net is made out of Nitex® nylon.
240,000.00 - ₹252,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Description Material
Availability Pricing
Wildco 16-C22 Hess Stream Bottom Sampler, 243 um Net 0549120 Mfr # 16-C22
COMPARE
Hess Stream Bottom Sampler, 243 um Net Stainless Steel, Nitex Nylon
₹252,000.00INR / Each
Wildco 16-C32 Hess Stream Bottom Sampler, 363 um Net 0549122 Mfr # 16-C32
COMPARE
Hess Stream Bottom Sampler, 363 um Net Stainless Steel, Nitex Nylon
₹240,000.00INR / Each
Wildco 16-C52 Hess Stream Bottom Sampler, 500 um (EPA) Net 0549124 Mfr # 16-C52
COMPARE
Hess Stream Bottom Sampler, 500 um (EPA) Net Stainless Steel, Nitex Nylon
₹252,000.00INR / Each