Wildco 16-C52 Hess Stream Bottom Sampler, 500 um (EPA) Net
Representative image only.
Wildco 16-C52 Hess Stream Bottom Sampler, 500 um (EPA) Net
Wildco | Mfr: 16-C52| Item: 0549124
Quantitative sampler for benthic invertebrates
₹252,000.00INR / Each
₹252,000.00INR / Each
Product Description
Key Features
  • Recommended for sampling shallow streams (<18") with gravel, cobble bottoms
  • Open base encloses area of 0.086 square meters
  • 3 month warranty
More About this Item
This 304 SS cylindrical sampler has open top and reversible handles for pushing and rotating into stream bottoms. Once in place, scrub the substrate inside so that specimens are carried by the current into the net. Includes Dolphin™ adapter and bucket for easy sample removal. Net is made out of Nitex® nylon.
3 Month Warranty
Specifications
  • MaterialStainless Steel, Nitex Nylon
  • DescriptionHess Stream Bottom Sampler, 500 um (EPA) Net