Wika Industrial Stainless Steel Gauges - Type 213.53
Accurately measure pressure in adverse conditions
  • Suitable for gaseous or liquid media
  • Vibration and shock resistant
  • NEMA 4X (IP 66) rated
These gauges feature a stainless steel case for protection in harsh environments. These gauges are supplied standard with a glycerine filled case. Suitable for environments compatible with copper alloy wetted parts where vibration or pressure pulsation occur and for gaseous or liquid media that will not obstruct the pressure system.
6,600.00 - ₹19,500.00INR / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Accuracy Dial Size (in) Max Pressure (PSI)
Availability Pricing
Wika 213.53 Liquid Filled Industrial Pressure Gauge, 0 to 3000 psi, 2.5" Dial, Lower 6843329 Mfr # 9692105
COMPARE
±2-1-2% full-scale 2.5 3000
₹6,600.00INR / Each
Wika 213.53 Liquid Filled Industrial Pressure Gauge, 30" Hg to 15 psi, 2.5" Dial, Back 6843354 Mfr # 4270898
COMPARE
±1.5% full-scale 2.5 15
₹10,800.00INR / Each
Wika 213.53 Liquid Filled Industrial Pressure Gauge, 0 to 30" Hg, 2.5" Dial, Lower 6843424 Mfr # 9693683
COMPARE
±2-1-2% full-scale 2.5 14.7
₹7,200.00INR / Each
Wika 213.53 Liquid Filled Industrial Pressure Gauge, 0 to 2000 psi, 4" Dial, Lower 6843425 Mfr # 9694183
COMPARE
±1% full scale 4 2000
₹15,300.00INR / Each
Wika 213.53 Liquid Filled Industrial Pressure Gauge, 0 to 200 psi, 4" Dial, Lower 6843427 Mfr # 9699134
COMPARE
±1% full scale 4 200
₹19,500.00INR / Each
Wika 213.53 Liquid Filled Industrial Pressure Gauge, 0 to 30" Hg, 4" Dial, Lower 6843478 Mfr # 9699053
COMPARE
±2-1-2% full-scale 4 14.7
₹14,800.00INR / Each
Wika 213.53 Liquid Filled Industrial Pressure Gauge, 0 to 60 psi, 4" Dial, Lower 6843479 Mfr # 9699117
COMPARE
±2-1-2% full-scale 4 60
₹14,300.00INR / Each
Wika 213.53 Liquid Filled Industrial Pressure Gauge, 30" Hg to 200 psi, 4" Dial, Lower 6843504 Mfr # 9699087
COMPARE
±1% full scale 4 200
₹14,800.00INR / Each
Wika 213.53 Liquid Filled Industrial Pressure Gauge, 0 to 300 psi, 4" Dial, Lower 6843505 Mfr # 9699142
COMPARE
±2-1-2% full-scale 4 300
₹14,300.00INR / Each
Showing 1- 9 of 9