Whirl Pak Bags without labels
Keep samples safe—easy-seal bags hold solids and liquids
  • Quick and easy sealing—flip bag around wire/tape and bend to seal
  • No more poking—puncture-proof tabs keep wire intact even after extensive use
All bags are sterilized after manufacturing using ethylene oxide gas at an independent ISO-certified facility. Exceptionally clear film and superior strength are achieved by using co-extrusion low-density and linear low-density, virgin polyethylene.
Capacity is measured in fluid ounces when the bag is closed, and tab is folded over three times. Volume and dimensions are approximate, bags should not be used at temperatures above 180° F (82° C). Bags can be frozen to any temperature, even liquid nitrogen
37,500.00 - ₹57,400.00INR / Box of 500
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (oz) Thickness (mil) Description
Availability Pricing
Whirl-Pak B00736WA Sterile sampling bag, 18 oz, Clear 0649920 Mfr # B00736
COMPARE
18 2.5 Sterile sampling bag, 18 oz, Clear
₹37,500.00INR / Box of 500
Whirl-Pak B01020WA Sterile sampling bag, 24 oz, Clear 0649930 Mfr # B01020
COMPARE
24 3 Sterile sampling bag, 24 oz, Clear
₹40,700.00INR / Box of 500
Whirl-Pak B00994WA Sterile sampling bag, 36 oz, Clear 0649940 Mfr # B00994
COMPARE
36 3 Sterile sampling bag, 36 oz, Clear
₹42,900.00INR / Box of 500
Whirl-Pak B01027WA Sterile sampling bag, 42 oz, Clear 0649950 Mfr # B01027
COMPARE
42 3 Sterile sampling bag, 42 oz, Clear
₹50,900.00INR / Box of 500
Whirl-Pak B01323WA Sterile Sampling Bag, 69 oz 500/bx 0649954 Mfr # B01323
COMPARE
69 3 Sterile Sampling Bag, 69 oz 500/bx
₹57,400.00INR / Box of 500