WHEATON® Vial Stoppers
Reliably seal vials
  • Choose from snap-on or straight plug vial stopper designs
Straight plug style stoppers provide maximum sealing for vials with straight wall glass finish. The straight wall interior finish is typically found in vials from European manufacturers. This style is not recommended for vials with a blow-back feature, which is a small ridge on the interior of the vial's neck.
Snap-on style stoppers provide maximum sealing for vials with a a blow-back glass finish, which is a small ridge on the interior of the vial's neck. The snap-on stopper locks into the blow-back ridge, providing a tight seal and keeping the stopper from backing out. This style is not recommended for vials with a straight-walled finish. The blow-back feature is typically found in vials from North American manufacturers.
32,200.00 - ₹60,900.00INR / Case of 1000
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Top Outside Diameter (OD) (mm) Description
Availability Pricing
DWK Life Sciences (Wheaton) W224100-081 Vial Stopper, Snap-On, Gray Chlorobutyl, 13 mm; 1000/Cs WW-08919-01 Mfr # W224100-081
COMPARE
Bromobutyl 13 Stopper, 13mm, Snap-On, Gray Butyl, 1000/CS
₹32,200.00INR / Case of 1000
DWK Life Sciences (Wheaton) W224100-181 Vial Stopper, Snap-On, Gray Chlorobutyl, 20 mm; 1000/Cs WW-08919-02 Mfr # W224100-181
COMPARE
Bromobutyl 20 Stopper, 20mm, Snap-On, Gray Butyl, 1000/CS
₹45,800.00INR / Case of 1000
DWK Life Sciences (Wheaton) W224100-400 Vial Stopper, Straight Plug, Gray Chlorobutyl, Ultra Pure, 13 mm; 1000/Cs WW-08919-04 Mfr # W224100-400
COMPARE
Bromobutyl 13 Stopper, 13mm, Straight Plug, Ultra Pure, Gray Butyl, 1000/CS
₹40,500.00INR / Case of 1000
DWK Life Sciences (Wheaton) W224100-405 Vial Stopper, Straight Plug, Gray Chlorobutyl, Ultra Pure, 20 mm; 1000/Cs WW-08919-05 Mfr # W224100-405
COMPARE
Bromobutyl 20 Stopper, 20mm, Straight Plug, Ultra Pure, Gray Butyl, 1000/CS
₹60,900.00INR / Case of 1000