WHEATON® High-Performance Electric Crimping and Decapping Tools
Reduce crimping and decapping time
  • Ergonomically designed to reduce strain
  • Use with flip-off caps
Quickly and easily crimp and decap laboratory vials and seals with aluminum or two-part aluminum/steel caps. These high-performance crimping tools are designed with interchangeable jaws—simply change the jaw set to the size you need. Fast and powerful, high-performance tools are strong enough for use with steel and magnetic caps. Crimping tools store up to 9 programs for different caps and cap sizes. Use with optional tool stand to reduce hand fatigue.
671,000.00 - ₹805,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
DWK Life Sciences (Wheaton) W225731-A High-Performance Crimper, 20 mm Jaw Set 9880011 Mfr # W225831-A
COMPARE
High-performance crimper with 20 mm jaw set
₹671,000.00INR / Each
DWK Life Sciences (Wheaton) W225732-A High-Performance Crimper and Decapper, 20 mm Jaw Set 9880012 Mfr # W225832-A
COMPARE
High-performance crimper and decapper with 20 mm jaw set
₹805,000.00INR / Each
DWK Life Sciences (Wheaton) W225733-A High-Performance Crimper, 20 mm Flip-Off Cap Jaw Set 9880013 Mfr # W225833-A
COMPARE
High-performance flip-off cap crimper and decapper with 20 mm jaw set
₹805,000.00INR / Each