DWK Life Sciences (Wheaton) W225732-A High-Performance Crimper and Decapper, 20 mm Jaw Set
Representative image only.
DWK Life Sciences (Wheaton) W225732-A High-Performance Crimper and Decapper, 20 mm Jaw Set
DWK Life Sciences (Wheaton) | Mfr: W225832-A| Item: 9880012
Reduce crimping and decapping time
₹805,000.00INR / Each
₹805,000.00INR / Each
Buy It Together
Current Item
DWK Life Sciences (Wheaton) W225732-A High-Performance Crimper and Decapper, 20 mm Jaw Set
DWK Life Sciences (Wheaton) W225732-A High-Performance Crimper and Decapper, 20 mm Jaw Set
9880012
₹805,000.00INR / Each
+
DWK Life Sciences (Wheaton) W225701 Crimping Tool Stand
9880024
₹75,500.00INR / Each
=
Total Price 880500.00 ADD 2 ITEMS TO CART
Product Description
Includes
A crimping jaw set, decapping jaw set, universal power supply, and power cord with plug.
Key Features
  • Ergonomically designed to reduce strain
  • Use with flip-off caps
  • 1 year warranty
More About this Item
Quickly and easily crimp and decap laboratory vials and seals with aluminum or two-part aluminum/steel caps. These high-performance crimping tools are designed with interchangeable jaws—simply change the jaw set to the size you need. Fast and powerful, high-performance tools are strong enough for use with steel and magnetic caps. Crimping tools store up to 9 programs for different caps and cap sizes. Use with optional tool stand to reduce hand fatigue.
This high-performance crimping tool comes with a crimping jaw set (98800-17) and a decapping jaw set (98800-20) for 20-mm caps.
1 Year Warranty
Specifications
  • DescriptionHigh-performance crimper and decapper with 20 mm jaw set