Vivaspin Centrifugal Concentrators with PES MWCO Membranes
Broad range of process alternatives for the filtration and concentration of biological samples
  • Disposable ultrafiltration devices concentrate and purify biological samples to concentrations over 100x in a single spin!
  • Concentrators feature twin vertical membranes—increased surface area provides faster filtration
  • Polyethersulfone membrane is excellent for most solutions when recovery is most important
The 500µl centrifugal filter units can be used in either swing out or fixed angle rotors accepting 2.2 ml centrifuge tubes. The vertical membrane design and thin channel filtration chamber minimizes membrane fouling and provides high speed concentrations, even with particle laden solutions.
34,100.00 - ₹37,900.00INR / Pkg of 25
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Hold-Up Volume (mL) Max Molecular Weight Cut Off (Mwco) (Dalton) Membrane
Availability Pricing
Sartorius Vivaspin 2 Centrifugal Concentrators, PES Membrane 5000 Da; 25/Pk 3622420 Mfr # VS0211
COMPARE
0.01 5000 Polyethersulfone
₹37,900.00INR / Pkg of 25
Sartorius Vivaspin 500 Centrifugal Concentrators, PES Membrane 5000 Da; 25/Pk 3622800 Mfr # VS0111
COMPARE
0.005 5000 Polyethersulfone
₹34,100.00INR / Pkg of 25
Sartorius Vivaspin 500 Centrifugal Concentrators, PES Membrane 30,000 Da; 25/Pk 3622804 Mfr # VS0121
COMPARE
0.005 30000 Polyethersulfone
₹34,100.00INR / Pkg of 25
Sartorius Vivaspin 500 Centrifugal Concentrators, PES Membrane 50,000 Da; 25/Pk 3622806 Mfr # VS0131
COMPARE
0.005 50000 Polyethersulfone
₹34,100.00INR / Pkg of 25
Sartorius Vivaspin 500 Centrifugal Concentrators, PES Membrane 100,000 Da; 25/Pk 3622808 Mfr # VS0141
COMPARE
0.005 100000 Polyethersulfone
₹34,100.00INR / Pkg of 25
Sartorius Vivaspin 500 Centrifugal Concentrators, PES Membrane 300,000 Da; 25/Pk 3622810 Mfr # VS0151
COMPARE
0.005 300000 Polyethersulfone
₹34,100.00INR / Pkg of 25
Sartorius Vivaspin 500 Centrifugal Concentrators, PES Membrane 1,000,000 Da; 25/Pk 3622812 Mfr # VS0161
COMPARE
0.005 1000000 Polyethersulfone
₹34,100.00INR / Pkg of 25
Showing 1- 7 of 7