Sartorius Vivaspin 500 Centrifugal Concentrators, PES Membrane 50,000 Da; 25/Pk

Representative image only.

Sartorius Vivaspin 500 Centrifugal Concentrators, PES Membrane 50,000 Da; 25/Pk

Sartorius | Mfr: VS0131| Item: 3622806

Broad range of process alternatives for the filtration and concentration of biological samples

₹34,100.00INR / Pkg of 25

₹34,100.00INR / Pkg of 25
Product Description

Key Features

  • Disposable ultrafiltration devices concentrate and purify biological samples to concentrations over 100x in a single spin!
  • Concentrators feature twin vertical membranes—increased surface area provides faster filtration
  • Polyethersulfone membrane is excellent for most solutions when recovery is most important

More About this Item

The 500µl centrifugal filter units can be used in either swing out or fixed angle rotors accepting 2.2 ml centrifuge tubes. The vertical membrane design and thin channel filtration chamber minimizes membrane fouling and provides high speed concentrations, even with particle laden solutions.

Specifications
  • MembranePolyethersulfone
  • Filtration Area (cm²)0.5
  • Hold-Up Volume (mL)0.005
  • Max Molecular Weight Cut Off (Mwco) (Dalton)50000
  • Length (cm)5
  • Width (cm)1.1
  • Max Volume Range (µL)0.6
  • Min Volume Range (mL)0.1
  • DescriptionCentrifugal Concentrators, PES Membrane 50,000 Da; 25/Pk
Reviews
Q&A