Vespel® Capillary Column Ferrules
Ideal for GC/MS interface applications
  • Manufactured of 60% Vespel and 40% Graphite
Vespel®/Graphite Ferrules are one-piece design requiring no back ferrule. Maximum temperature: 400°C. Recommended for GC/MS interface applications. Reusable – does not seize into column.
6,400.00INR / Pkg of 10
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
VESPel Graphite Capillary Column Ferrules, 60% Vespel and 40% Graphite, 1/16" to 0.4 mm; 10/Pk WW-01998-60 Mfr # 213104
COMPARE
Graphite Capillary Column Ferrules, 1/16\" to 0.4 mm
₹6,400.00INR / Pkg of 10
VESPel Graphite Capillary Column Ferrules, 60% Vespel and 40% Graphite, 1/16" to 0.5 mm; 10/Pk WW-01998-62 Mfr # 213105
COMPARE
Graphite Capillary Column Ferrules, 1/16\" to 0.5 mm
₹6,400.00INR / Pkg of 10
VESPel Graphite Capillary Column Ferrules, 60% Vespel and 40% Graphite, 1/16" to 0.8 mm; 10/Pk WW-01998-64 Mfr # 213108
COMPARE
Graphite Capillary Column Ferrules, 1/16\" to 0.8 mm
₹6,400.00INR / Pkg of 10