Vanguard Safety C37B6 Neoprene Coated Latex Flock Lined Gloves, 28 mil, 12" Cuff, Blue/Yellow
Two-tone color indicates glove wear over time
  • Non-slip honeycomb surface ensures secure grip
  • Protects against most chemicals
  • Complies with FFDCA food handling requirements 21 CFR, Parts 170-199
These reusable neoprene coated latex flock lined gloves feature a multi-layer design to protect you from most chemicals. The flock-lining serves as an extra layer of protection, provides increased comfort, and easy donning. Honeycomb non-slip surface offers secure handling in dry, wet, and oily conditions. The gloves should be replaced when the yellow color of the glove shows through the blue.
The 28-mil thick gloves are ideal for use in industrial, manufacturing, janitorial, automotive, chemical processing, food service, and food processing industries.
59,100.00INR / Case of 144
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Size Thickness (mil)
Availability Pricing
Vanguard Safety Neoprene Coated Latex Flock Lined Gloves, 28 mil, 12" Cuff, Blue/Yellow,  Medium; 144/CS 7890677 Mfr # C37B68
COMPARE
Latex Medium 28
₹59,100.00INR / Case of 144
Vanguard Safety Neoprene Coated Latex Flock Lined Gloves, 28 mil, 12" Cuff, Blue/Yellow,  Laege; 144/CS 7890678 Mfr # C37B69
COMPARE
Latex Large 28
₹59,100.00INR / Case of 144
Vanguard Safety Neoprene Coated Latex Flock Lined Gloves, 28 mil, 12" Cuff, Blue/Yellow,  Extra-Large; 144/CS 7890679 Mfr # C37B610
COMPARE
Latex Extra Large 28
₹59,100.00INR / Case of 144