Vanguard Safety Neoprene Coated Latex Flock Lined Gloves, 28 mil, 12" Cuff, Blue/Yellow, Medium; 144/CS
Representative image only.
Vanguard Safety Neoprene Coated Latex Flock Lined Gloves, 28 mil, 12" Cuff, Blue/Yellow, Medium; 144/CS
Vanguard Safety | Mfr: C37B68| Item: 7890677
Two-tone color indicates glove wear over time
₹59,100.00INR / Case of 144
₹59,100.00INR / Case of 144
Product Description
Key Features
  • Non-slip honeycomb surface ensures secure grip
  • Protects against most chemicals
  • Complies with FFDCA food handling requirements 21 CFR, Parts 170-199
  • 2 month warranty
More About this Item
These reusable neoprene coated latex flock lined gloves feature a multi-layer design to protect you from most chemicals. The flock-lining serves as an extra layer of protection, provides increased comfort, and easy donning. Honeycomb non-slip surface offers secure handling in dry, wet, and oily conditions. The gloves should be replaced when the yellow color of the glove shows through the blue.
The 28-mil thick gloves are ideal for use in industrial, manufacturing, janitorial, automotive, chemical processing, food service, and food processing industries.
2 Month Warranty
Specifications
  • Thickness (mil)28
  • MaterialLatex
  • SizeMedium
  • ColorBlue/Yellow
  • LiningFlock
  • GripHoneycomb
  • Cuff12" Straight
  • ComplianceFFDCA compliant
  • SterileNo
  • DescriptionNeoprene Coated Latex Flock Lined Gloves, 28 mil, 12" Cuff, Blue/Yellow, Medium; 144/CS
  • Warranty2 months