Vanguard Safety C25B3 Latex Flock Lined Gloves, 16 mil, 12” Cuff, Blue
Protects against acids, bases, and solvents
  • Non-slip honeycomb surface ensures secure grip
  • Beaded cuff
  • Complies with FFDCA food handling requirements 21 CFR, Parts 170-199
These reusable flock lined gloves offer you increased comfort and easy donning. A beaded cuff prevents potential product contamination while a non-slip honeycomb surface delivers a secure grip. The form-fitting latex gloves are snag, puncture, and abrasion resistant. Ideal for use in food processing, food service, janitorial, industrial, manufacturing, and chemical processing industries.
27,500.00 - ₹29,800.00INR / Case of 144
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Size Thickness (mil)
Availability Pricing
Vanguard Safety Latex Flock Lined Gloves, 16 mil, 13" Cuff, Blue, Medium; 144/CS 7890662 Mfr # C25B38
COMPARE
Latex Medium 16
₹27,500.00INR / Case of 144
Vanguard Safety Latex Flock Lined Gloves, 16 mil, 13" Cuff, Blue, Large; 144/CS 7890663 Mfr # C25B39
COMPARE
Latex Large 16
₹27,500.00INR / Case of 144
Vanguard Safety Latex Flock Lined Gloves, 16 mil, 13" Cuff, Blue, Extra-Large; 144/CS 7890664 Mfr # C25B310
COMPARE
Latex Extra Large 16
₹29,800.00INR / Case of 144
Vanguard Safety Latex Flock Lined Gloves, 16 mil, 13" Cuff, Blue, 2X-Large; 144/CS 7890665 Mfr # C25B311
COMPARE
Latex 2X Large 16
₹28,400.00INR / Case of 144