Vanguard Safety Latex Flock Lined Gloves, 16 mil, 13" Cuff, Blue, Extra-Large; 144/CS
Representative image only.
Vanguard Safety Latex Flock Lined Gloves, 16 mil, 13" Cuff, Blue, Extra-Large; 144/CS
Vanguard Safety | Mfr: C25B310| Item: 7890664
Protects against acids, bases, and solvents
₹29,800.00INR / Case of 144
₹29,800.00INR / Case of 144
Product Description
Key Features
  • Non-slip honeycomb surface ensures secure grip
  • Beaded cuff
  • Complies with FFDCA food handling requirements 21 CFR, Parts 170-199
  • 2 month warranty
More About this Item
These reusable flock lined gloves offer you increased comfort and easy donning. A beaded cuff prevents potential product contamination while a non-slip honeycomb surface delivers a secure grip. The form-fitting latex gloves are snag, puncture, and abrasion resistant. Ideal for use in food processing, food service, janitorial, industrial, manufacturing, and chemical processing industries.
2 Month Warranty
Specifications
  • MaterialLatex
  • Thickness (mil)16
  • SizeExtra Large
  • ColorBlue
  • LiningFlock
  • GripHoneycomb
  • Cuff13" Rolled
  • ComplianceFFDCA compliant
  • SterileNo
  • DescriptionLatex Flock Lined Gloves, 16 mil, 13" Cuff, Blue, Extra-Large; 144/CS
  • Warranty2 months