Vanguard Safety C23A3 Latex Unlined Gloves, 13 mil, 13" Cuff, Blue
Provides dexterity in wet and dry environments
  • Double chlorinated for easy donning
  • Non-slip fish scale surface ensures secure grip
  • Beaded cuff
  • Complies with FFDCA food handling requirements 21 CFR, Parts 170-199
These reusable gloves are unlined and offer a beaded cuff to reduce potential product contamination. A non-slip fish scale surface delivers a secure grip. Form-fitting latex provides excellent dexterity in wet and dry environments. Ideal for use in food processing, food service, janitorial, industrial, manufacturing, and chemical processing industries.
29,800.00INR / Case of 144
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Size Thickness (mil)
Availability Pricing
Vanguard Safety Latex Unlined Gloves, 13 mil, 13" Cuff, Blue, Medium; 144/CS 7890655 Mfr # C23A38
COMPARE
Latex Medium 13
₹29,800.00INR / Case of 144
Vanguard Safety Latex Unlined Gloves, 13 mil, 13" Cuff, Blue, Large; 144/CS 7890656 Mfr # C23A39
COMPARE
Latex Large 13
₹29,800.00INR / Case of 144
Vanguard Safety Latex Unlined Gloves, 13 mil, 13" Cuff, Blue, Extra-Large; 144/CS 7890657 Mfr # C23A310
COMPARE
Latex Extra Large 13
₹29,800.00INR / Case of 144