Vanguard Safety Latex Unlined Gloves, 13 mil, 13" Cuff, Blue, Extra-Large; 144/CS
Representative image only.
Vanguard Safety Latex Unlined Gloves, 13 mil, 13" Cuff, Blue, Extra-Large; 144/CS
Vanguard Safety | Mfr: C23A310| Item: 7890657
Provides dexterity in wet and dry environments
₹29,800.00INR / Case of 144
₹29,800.00INR / Case of 144
Product Description
Key Features
  • Double chlorinated for easy donning
  • Non-slip fish scale surface ensures secure grip
  • Beaded cuff
  • Complies with FFDCA food handling requirements 21 CFR, Parts 170-199
  • 2 month warranty
More About this Item
These reusable gloves are unlined and offer a beaded cuff to reduce potential product contamination. A non-slip fish scale surface delivers a secure grip. Form-fitting latex provides excellent dexterity in wet and dry environments. Ideal for use in food processing, food service, janitorial, industrial, manufacturing, and chemical processing industries.
2 Month Warranty
Specifications
  • MaterialLatex
  • Thickness (mil)13
  • SizeExtra Large
  • ColorBlue
  • LiningUnlined
  • GripFish Scale
  • Cuff13" Rolled
  • ComplianceFFDCA compliant
  • SterileNo
  • DescriptionLatex Unlined Gloves, 13 mil, 13" Cuff, Blue, Extra-Large; 144/CS
  • Warranty2 months