Vanguard Safety A1DH1 Powder-Free Nitrile Exam Gloves, 8.7 mil, Blue
Protects against chemicals, oils, and blood borne pathogens
  • Fully texture surface for secure grip
  • Powder-free
  • Chemotherapy rated
  • Medical grade – ASTM D6319
  • Complies with FFDCA food handling requirements 21 CFR, Parts 170-199
  • Ambidextrous
Protect your wrists and forearms with a 12” extended cuff. The 8.7-mil thick nitrile gloves provide excellent puncture and abrasion resistance along with a fully textured surface for a secure grip. The gloves are powder-free to reduce skin irritation, tested for use against Fentanyl as per ADTM D6978-05, and latex free. Ideal for use in chemotherapy, automotive, food processing, janitorial, food service, healthcare, industrial, manufacturing, agriculture, chemical processing, vet, and laboratory industries.
22,800.00INR / Case of 500
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (in) Material Size
Availability Pricing
Vanguard Safety Powder-Free Nitrile Chemo Exam Gloves, 8.7 mil, 12", Blue, Small; 500/CS 7890633 Mfr # A1DH11
COMPARE
12 Nitrile Small
₹22,800.00INR / Case of 500
Vanguard Safety Powder-Free Nitrile Chemo Exam Gloves, 8.7 mil, Blue, Medium; 500/CS 7890634 Mfr # A1DH12
COMPARE
12 Nitrile Medium
₹22,800.00INR / Case of 500
Vanguard Safety Powder-Free Nitrile Chemo Exam Gloves, 8.7 mil, Blue, Large; 500/CS 7890635 Mfr # A1DH13
COMPARE
12 Nitrile Large
₹22,800.00INR / Case of 500
Vanguard Safety Powder-Free Nitrile Chemo Exam Gloves, 8.7 mil, Blue, Extra-Large; 500/CS 7890636 Mfr # A1DH14
COMPARE
12 Nitrile X-Large
₹22,800.00INR / Case of 500