Vanguard Safety Powder-Free Nitrile Chemo Exam Gloves, 8.7 mil, Blue, Medium; 500/CS
Representative image only.
Vanguard Safety Powder-Free Nitrile Chemo Exam Gloves, 8.7 mil, Blue, Medium; 500/CS
Vanguard Safety | Mfr: A1DH12| Item: 7890634
Protects against chemicals, oils, and blood borne pathogens
₹22,800.00INR / Case of 500
₹22,800.00INR / Case of 500
Product Description
Key Features
  • Fully texture surface for secure grip
  • Powder-free
  • Chemotherapy rated
  • Medical grade – ASTM D6319
  • Complies with FFDCA food handling requirements 21 CFR, Parts 170-199
  • Ambidextrous
  • 2 month warranty
More About this Item
Protect your wrists and forearms with a 12” extended cuff. The 8.7-mil thick nitrile gloves provide excellent puncture and abrasion resistance along with a fully textured surface for a secure grip. The gloves are powder-free to reduce skin irritation, tested for use against Fentanyl as per ADTM D6978-05, and latex free. Ideal for use in chemotherapy, automotive, food processing, janitorial, food service, healthcare, industrial, manufacturing, agriculture, chemical processing, vet, and laboratory industries.
2 Month Warranty
Specifications
  • Thickness (mil)8.7
  • MaterialNitrile
  • SizeMedium
  • Length (in)12
  • Length (cm)29
  • ColorBlue
  • GripTextured
  • ComplianceFDA approved
  • SterileNo
  • PowderedNo
  • Palm Thickness (mil)7.8
  • DescriptionPowder-Free Nitrile Chemo Exam Gloves, 8.7 mil, Blue, Medium; 500/CS
  • Warranty2 months