Value Plastics Male Luer to 500 Series Barb
Luer tapers that are constructed to meet ISO 594-1 and 594-2 standards
  • Rotating locks minimize twisting and kinking of tubing during positioning
  • Tapered cone seals are reliable and uncomplicated
  • Facilitate single-use configurations that minimize downtime
These luer fittings come in many configurations and several materials, joining tubing from 1/16” (1.6 mm) ID to 1/4” (6.4 mm) ID. The 500 series barb is composed of blended radii giving it a distinct look. Each barb is designed to accommodate a range of tube sizes and tube types. Consideration should be given to flow restriction, pressure capability, and the force required for assembly of the tube. Because the fittings are made to ISO standards that are well understood globally, it ensures proper interconnections and often removes the need to retrofit*.
Options include polycarbonate or polypropylene resin.
Polycarbonate is transparent and features high impact, chemical, UV, and electrical resistance. Sterilization is only recommended via ethylene oxide (EtO). Gamma radiation up to 10 Mrads is possible with minor loss of physical properties – it will discolor to a yellow-green hue. Autoclave is not recommended and may crack the fitting.
Polypropylene is clarified and produced without any animal derived components or phthalates. It can be sterilized using gamma radiation, Ethylene oxide (EtO), E-beam, or autoclave.
2,700.00 - ₹2,900.00INR / Pkg of 25
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 2) (in) Fitting Material Luer Connection Type (Port 1)
Availability Pricing
Value Plastics Luer to Hose Barb Fitting, Straight Adapter, Polypropylene, Male Luer x 500 Series Barb 1/8" ID; 25/Pk 5012012 Mfr # MLRL013-6005
COMPARE
1/8 Polypropylene Male Luer
₹2,700.00INR / Pkg of 25
Value Plastics Luer to Hose Barb Fitting, Straight Adapter, Polycarbonate, Male Luer x 500 Series Barb 1/4" ID; 25/Pk 5012013 Mfr # MLRL055-9
COMPARE
1/4 Polycarbonate Male Luer
₹2,900.00INR / Pkg of 25