Value Plastics Luer to Hose Barb Fitting, Straight Adapter, Polycarbonate, Male Luer x 500 Series Barb 1/4" ID; 25/Pk
Representative image only.
Value Plastics Luer to Hose Barb Fitting, Straight Adapter, Polycarbonate, Male Luer x 500 Series Barb 1/4" ID; 25/Pk
Value Plastics | Mfr: MLRL055-9| Item: 5012013
Luer tapers that are constructed to meet ISO 594-1 and 594-2 standards
₹2,900.00INR / Pkg of 25
₹2,900.00INR / Pkg of 25
Product Description
Key Features
  • Rotating locks minimize twisting and kinking of tubing during positioning
  • Tapered cone seals are reliable and uncomplicated
  • Facilitate single-use configurations that minimize downtime
More About this Item
These luer fittings come in many configurations and several materials, joining tubing from 1/16” (1.6 mm) ID to 1/4” (6.4 mm) ID. The 500 series barb is composed of blended radii giving it a distinct look. Each barb is designed to accommodate a range of tube sizes and tube types. Consideration should be given to flow restriction, pressure capability, and the force required for assembly of the tube. Because the fittings are made to ISO standards that are well understood globally, it ensures proper interconnections and often removes the need to retrofit*.
Options include polycarbonate or polypropylene resin.
Polycarbonate is transparent and features high impact, chemical, UV, and electrical resistance. Sterilization is only recommended via ethylene oxide (EtO). Gamma radiation up to 10 Mrads is possible with minor loss of physical properties – it will discolor to a yellow-green hue. Autoclave is not recommended and may crack the fitting.
Polypropylene is clarified and produced without any animal derived components or phthalates. It can be sterilized using gamma radiation, Ethylene oxide (EtO), E-beam, or autoclave.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Certificate of Compliance
Specifications
  • Fitting MaterialPolycarbonate
  • Connection TypeMale Luer x Hose Barb
  • Number Of Ports2
  • Barb Size / Tubing Id (Port 2) (in)1/4
  • Flow PatternStraight
  • Regulatory ApprovalsNon-DEHP;Non-Phthalate;FDA;RoHS
  • Luer Connection Type (Port 1)Male Luer
  • DescriptionLuer to Hose Barb Fitting, Straight Adapter, Polycarbonate, Male Luer x 500 Series Barb 1/4" ID; 25/Pk