Value Plastics Male Luer Slip to Plug Adapters
Seal luer ports with a secure and leak-proof connection
  • Luer fittings allow for a simple yet secure connection
  • Ideal for capping off fluid transfer system
  • Quickly connect and disconnect with slip luer design
These Value Plastics Male Luer Slip to Plug Adapters are designed to easily plug female luer ports. Luer fittings ensure a tight and reliable connection while allowing for quick and easy assembly and disassembly without the need for tools. Plugs closed at the grip feature a sealing mechanism located closer to the body of the plug, where it connects with the mating female luer fitting. This design may be preferred in applications where the tip of the fitting needs to remain open or where the focus is on sealing at the base rather than the tip. Adapters are available in various materials to accommodate various applications and compliance requirements.
2,400.00 - ₹3,200.00INR / Pkg of 25
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Fitting Material Regulatory Approvals Description
Availability Pricing
Value Plastics Luer Fitting, Straight Adapter, White Nylon, Male Luer Slip x Plug, Closed at Grip; 25/Pk 5012081 Mfr # MTLSP-1
COMPARE
Nylon Non-DEHP Luer Fitting, Straight Adapter, White Nylon, Male Luer Slip x Plug, Closed at Grip; 25/Pk
₹2,400.00INR / Pkg of 25
Value Plastics Luer Fitting, Straight Adapter, Polypropylene, Male Luer Slip x Plug, Closed at Grip; 25/Pk 5012082 Mfr # MTLSP-6005
COMPARE
Polypropylene RoHS Luer Fitting, Straight Adapter, Polypropylene, Male Luer Slip x Plug, Closed at Grip; 25/Pk
₹3,200.00INR / Pkg of 25