Value Plastics Luer Fitting, Straight Adapter, White Nylon, Male Luer Slip x Plug, Closed at Grip; 25/Pk
Representative image only.
Value Plastics Luer Fitting, Straight Adapter, White Nylon, Male Luer Slip x Plug, Closed at Grip; 25/Pk
Value Plastics | Mfr: MTLSP-1| Item: 5012081
Seal luer ports with a secure and leak-proof connection
₹2,400.00INR / Pkg of 25
₹2,400.00INR / Pkg of 25
Product Description
Key Features
  • Luer fittings allow for a simple yet secure connection
  • Ideal for capping off fluid transfer system
  • Quickly connect and disconnect with slip luer design
  • Constructed of durable chemical resistance nylon
More About this Item
These Value Plastics Male Luer Slip to Plug Adapters are designed to easily plug female luer ports. Luer fittings ensure a tight and reliable connection while allowing for quick and easy assembly and disassembly without the need for tools. Plugs closed at the grip feature a sealing mechanism located closer to the body of the plug, where it connects with the mating female luer fitting. This design may be preferred in applications where the tip of the fitting needs to remain open or where the focus is on sealing at the base rather than the tip. Adapters are available in various materials to accommodate various applications and compliance requirements.
White nylon is suitable for Gamma radiation, ethylene oxide (EtO), and autoclave sterilization.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Certificate of Compliance
Specifications
  • Fitting MaterialNylon
  • Width (InchShort)3/8
  • Height (InchShort)9/16
  • Number Of Ports1
  • Fitting Width (in)3/8
  • Regulatory ApprovalsNon-DEHP;RoHS;Non-Phthalate
  • Luer Connection Type (Port 1)Male Luer Slip
  • Luer Connection Type (Port 2)Plug, Closed at Grip
  • DescriptionLuer Fitting, Straight Adapter, White Nylon, Male Luer Slip x Plug, Closed at Grip; 25/Pk