Value Plastics Female Luer Thread Style Caps
Seal luer ports with a secure and leak-proof connection
  • Luer fittings allow for a simple yet secure twist-lock connection
  • Ideal for capping off fluid transfer system
  • Thread design provides a secure closure—preventing contamination
These Value Plastics Female Luer Thread Style Caps are designed to easily plug male luer ports. Luer fittings ensure a tight and reliable connection while allowing for quick and easy assembly and disassembly without the need for tools. These adapters are available in various materials to accommodate various applications and compliance requirements.
2,400.00 - ₹2,900.00INR / Pkg of 25
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Fitting Material Regulatory Approvals Description
Availability Pricing
Value Plastics Luer Fitting, Clear Polycarbonate, Female Luer Thread Cap; 25/Pk 5012041 Mfr # FTLLP-9
COMPARE
Polycarbonate FDA Luer Fitting, Clear Polycarbonate, Female Luer Thread Cap; 25/Pk
₹2,900.00INR / Pkg of 25
Value Plastics Luer Fitting, White Nylon, Female Luer Thread Cap; 25/Pk 5012053 Mfr # FTLLP-1
COMPARE
Nylon Non-DEHP Luer Fitting, White Nylon, Female Luer Thread Cap; 25/Pk
₹2,400.00INR / Pkg of 25