Value Plastics Luer Fitting, Clear Polycarbonate, Female Luer Thread Cap; 25/Pk
Representative image only.
Value Plastics Luer Fitting, Clear Polycarbonate, Female Luer Thread Cap; 25/Pk
Value Plastics | Mfr: FTLLP-9| Item: 5012041
Seal luer ports with a secure and leak-proof connection
₹2,900.00INR / Pkg of 25
₹2,900.00INR / Pkg of 25
Buy It Together
Current Item
Value Plastics Luer Fitting, Clear Polycarbonate, Female Luer Thread Cap; 25/Pk
Value Plastics Luer Fitting, Clear Polycarbonate, Female Luer Thread Cap; 25/Pk
5012041
₹2,900.00INR / Pkg of 25
+ =
Total Price 4300.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Luer fittings allow for a simple yet secure twist-lock connection
  • Ideal for capping off fluid transfer system
  • Thread design provides a secure closure—preventing contamination
  • Constructed of high temperature resistant polycarbonate
More About this Item
These Value Plastics Female Luer Thread Style Caps are designed to easily plug male luer ports. Luer fittings ensure a tight and reliable connection while allowing for quick and easy assembly and disassembly without the need for tools. These adapters are available in various materials to accommodate various applications and compliance requirements.
Clear polycarbonate (PC) is suitable for sterilization via ethylene oxide (EtO).
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Certificate of Compliance
Specifications
  • Fitting MaterialPolycarbonate
  • Width (InchShort)11/32
  • Height (InchShort)7/16
  • Number Of Ports1
  • Fitting Width (in)11/32
  • Regulatory ApprovalsFDA;Non-Phthalate;RoHS;Non-DEHP
  • Luer Connection Type (Port 1)Female Luer Thread
  • Luer Connection Type (Port 2)Cap
  • DescriptionLuer Fitting, Clear Polycarbonate, Female Luer Thread Cap; 25/Pk