UVX Digital Ultraviolet Intensity Meter
With interchangeable NIST-traceable sensors
This radiometer is used to monitor the useful life of any UV light source by measuring lamp irradiance. It operates with a 9 V battery for up to 120 hours. Select three intensity ranges: 0 to 20 mW/cm2, 0 to 2000 µW/cm2, and 0 to 200 µW/cm2. Optional 10:1 attenuator extends the reading range up to 200 mW/cm2.
Choose one of the three interchangeable sensors for measuring 254 nm, 302 nm, or 365 nm ultraviolet wavelengths (sold separately). All sensors are calibrated with NIST traceability, and are ±5% accurate. Sensors must be purchased separately. The 3-ft cable length is ideal for measuring UV in remote locations.
124,000.00 - ₹280,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Analytik Jena 97-0016-01 Ultraviolet Intensity Radiometer Sensor, 254 nm 9756120 Mfr # 97-0016-01
COMPARE
UVX Ultraviolet Intensity Radiometer Sensor, 254 nm
₹124,000.00INR / Each
Analytik Jena 97-0015-02(UVX) Digital Ultraviolet Intensity Meter/Radiometer without Sensor 9765110 Mfr # 97-0015-02(UVX)
COMPARE
Digital Ultraviolet Intensity Meter/Radiometer without Sensor
₹280,000.00INR / Each