Analytik Jena 97-0015-02(UVX) Digital Ultraviolet Intensity Meter/Radiometer without Sensor

Representative image only.

Analytik Jena 97-0015-02(UVX) Digital Ultraviolet Intensity Meter/Radiometer without Sensor

Analytik Jena | Mfr: 97-0015-02(UVX)| Item: 9765110

With interchangeable NIST-traceable sensors

₹280,000.00INR / Each

₹280,000.00INR / Each
Product Description

Key Features

  • 1 year warranty

More About this Item

This radiometer is used to monitor the useful life of any UV light source by measuring lamp irradiance. It operates with a 9 V battery for up to 120 hours. Select three intensity ranges: 0 to 20 mW/cm2, 0 to 2000 µW/cm2, and 0 to 200 µW/cm2. Optional 10:1 attenuator extends the reading range up to 200 mW/cm2.
Choose one of the three interchangeable sensors for measuring 254 nm, 302 nm, or 365 nm ultraviolet wavelengths (sold separately). All sensors are calibrated with NIST traceability, and are ±5% accurate. Sensors must be purchased separately. The 3-ft cable length is ideal for measuring UV in remote locations.

1 Year WarrantyNIST Traceable Calibration
Specifications
  • Width (in)3 3/8
  • Height (InchShort)2
  • Depth (in)6 1/2
  • Width (cm)8.6
  • Height (cm)5.1
  • Depth (cm)16.5
  • Display TypeLCD, 3.5-digit
  • Battery1 9V (not included)
  • Intensity (µW/cm2)200/2000
  • Intensity (mW/cm2)20
  • DescriptionDigital Ultraviolet Intensity Meter/Radiometer without Sensor
  • Warranty1 year
Reviews
Q&A