US Industrial Multi-purpose Digital Gas Detectors with Two-Stage Alarm
Get readings of an area even from a distance
  • Continuous 24 hour monitoring Compact, durable units Explosion-proof housing for sensor Audio/Visual alarms Malfunction LED and test/checking Low, High alarm set switches Infrequent maintenance and calibration required Remote sensor reliable to 600 feet from controller or longer Relay contacts and recorder terminals Big 0.30" red LED digital display
Remote sensors provides open space monitoring ability.
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Gases Measured
Availability Pricing
US Industrial ET6200R/2AL/D Gas Detector w/2-Stage Alarm, ETO; Wall-Mt/Remote Sensor WW-10109-75 Mfr # ET6200R/2AL/D
COMPARE
Ethylene Oxide
Call For Price
US Industrial 7200R/2AL/D Gas Detector w/2-Stage Alarm, H2; Wall-Mt/Remote Sensor WW-10109-77 Mfr # 7200R/2AL/D
COMPARE
Hydrogen
Call For Price
US Industrial 8100R/2ALD Gas Detector w/2-Stage Alarm, LEL; Wall-Mt/Remote Sensor WW-10109-79 Mfr # 8100R/2ALD
COMPARE
Combustibles (methane)