United Scientific Tall-Form Glass Beakers

Save space with tall-form beaker
  • Large white printed graduations and a marking area
  • Constructed of durable borosilicate glass
United Scientific brand beakers are made from Type l borosilicate glass. Beakers feature heavy and uniform wall thickness for enhanced durability. Spout and beaded top is designed for easy pouring.
8,100.00 - ₹17,900.00INR / Pkg of 12

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Material
Availability Pricing
Borosil BG1060-100 Tall-Form Glass Beaker; 100 mL, 12/Pk 6100073 Mfr # BG1060-100
COMPARE
100 Glass
₹10,400.00INR / Pkg of 12
Borosil BG1060-250 Tall-Form Glass Beaker; 250 mL, 12/Pk 6100075 Mfr # BG1060-250
COMPARE
250 Glass
₹10,500.00INR / Pkg of 12
Borosil BG1060-500 Tall-Form Glass Beaker; 500 mL, 6/Pk 6100077 Mfr # BG1060-500
COMPARE
500 Glass
₹8,100.00INR / Pkg of 6
Borosil BG1060-1000 Tall-Form Glass Beaker; 1000 mL, 6/Pk 6100079 Mfr # BG1060-1000
COMPARE
1000 Glass
₹17,900.00INR / Pkg of 6
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat