United Scientific Supplies Liebig and Graham Condensers
Safely distill samples without complete evaporation
  • Made from 3.3 borosilicate glass for greater durability
  • Designed with 24/40 joint with screw thread connector
  • Ideal for botanical distillation or fragrance extraction
Both the Graham and Liebig condensers are manufactured from high-quality 3.3 borosilicate glass and feature a 24/40 flask joints with a screw thread connector. It also includes a condenser tube and two 1/4" hose nozzles, one for water in and one for water out.
Both condenser types feature an inner tube which contains the distillation sample and an outer tube for the coolant, or water. The water cools hot vapors within the inner tube to transfer the gaseous vapors to a liquid phase for condensation.
The Liebig condenser has a straight inner tube which is better suited for basic laboratory applications, or applications in which the substance could easily get clogged inside the inner tube.
The Graham condenser has a coiled inner tube, providing more surface area to cool more of your distillation substance at a time.
9,600.00 - ₹20,100.00INR / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Body Length (in) Condenser Type Description
Availability Pricing
United Scientific Supplies Graham Condenser, 24/40 Joint; 200 mm 3450953 Mfr # CNG200
COMPARE
7.87 Graham Graham Condenser, 24/40 Joint; 200 mm
₹12,800.00INR / Each
United Scientific Supplies Graham Condenser, 24/40 Joint; 300 mm 3450954 Mfr # CNG300
COMPARE
11.8 Graham Graham Condenser, 24/40 Joint; 300 mm
₹16,100.00INR / Each
United Scientific Supplies Graham Condenser, 24/40 Joint; 400 mm 3450955 Mfr # CNG400
COMPARE
15.75 Graham Graham Condenser, 24/40 Joint; 400 mm
₹20,100.00INR / Each
United Scientific Supplies Graham Condenser, 24/40 Joint; 500 mm 3450956 Mfr # CNG500
COMPARE
19.7 Graham Graham Condenser, 24/40 Joint; 500 mm
₹18,400.00INR / Each
United Scientific Supplies Liebig Condenser, 24/40 Joint; 200 mm 3450957 Mfr # CNL200
COMPARE
7.87 Liebig Liebig Condenser, 24/40 Joint; 200 mm
₹9,600.00INR / Each
United Scientific Supplies Liebig Condenser, 24/40 Joint; 300 mm 3450958 Mfr # CNL300
COMPARE
11.8 Liebig Liebig Condenser, 24/40 Joint; 300 mm
₹12,800.00INR / Each
United Scientific Supplies Liebig Condenser, 24/40 Joint; 400 mm 3450959 Mfr # CNL400
COMPARE
15.75 Liebig Liebig Condenser, 24/40 Joint; 400 mm
₹12,000.00INR / Each
United Scientific Supplies Liebig Condenser, 24/40 Joint; 500 mm 3450960 Mfr # CNL500
COMPARE
19.7 Liebig Liebig Condenser, 24/40 Joint; 500 mm
₹13,800.00INR / Each
Showing 1- 8 of 8