United Scientific™ Red Spirit-Filled Thermometers
Kerosene-based column is safer than mercury
  • Partial or total immersion to fit your unique application
  • Kersone-based red spirit is safer to use
  • Include no-roll sleeves to prevent accidental breakage
Red spirit-filled thermometers provide increased safety with economic pricing. These thermometers have 6 to 7 mm outer diameters and the bulbs have equal or smaller diameters than the stems. All thermometers include a no-roll sleeve to stop thermometers rolling off of lab benches.
Partial immersion thermometers are useful if you have a shallow container. These thermometers have a small indicator ring that denotes the depth to which the thermometer should be submerged to obtain an accurate temperature reading.
Total immersion thermometers need to be submerged deep enough so the desired temperature is at the meniscus.
930.00 - ₹1,200.00INR / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
Compareitem specifications
Overall Length (InchShort) Thermometer Fill Description
Availability Pricing
United Scientific Supplies Red Spirit-Filled Thermometer, 12" Length, Total Immersion, -20 to 110°C 7890952 Mfr # THTC01
COMPARE
12 Red spirit Red Spirit-Filled Thermometer, 12" Length, Total Immersion, -20 to 110°C
₹1,100.00INR / Each
United Scientific Supplies Red Spirit-Filled Thermometer, 12" Length, Total Immersion, -20 to 150°C 7890953 Mfr # THTC02
COMPARE
12 Red spirit Red Spirit-Filled Thermometer, 12" Length, Total Immersion, -20 to 150°C
₹1,100.00INR / Each
United Scientific Supplies Red Spirit-Filled Thermometer, 12" Length, Partial Immersion, -20 to 110°C 7890954 Mfr # THPC01
COMPARE
12 Red spirit Red Spirit-Filled Thermometer, 12" Length, Partial Immersion, -20 to 110°C
₹1,200.00INR / Each
United Scientific Supplies Red Spirit-Filled Thermometer, 12" Length, Partial Immersion, -10 to 110°C 7890955 Mfr # THPC02
COMPARE
12 Red spirit Red Spirit-Filled Thermometer, 12" Length, Partial Immersion, -10 to 110°C
₹1,200.00INR / Each
United Scientific Supplies Red Spirit-Filled Thermometer, 12" Length, Total Immersion, -20 to 110°C / 0 to 230°F 7890956 Mfr # THTCF1
COMPARE
12 Red spirit Red Spirit-Filled Thermometer, 12" Length, Total Immersion, -20 to 110°C / 0 to 230°F
₹1,200.00INR / Each
United Scientific Supplies Red Spirit-Filled Thermometer, 12" Length, Total Immersion, -20 to 150°C / 0 to 300°F 7890957 Mfr # THTCF2
COMPARE
12 Red spirit Red Spirit-Filled Thermometer, 12" Length, Total Immersion, -20 to 150°C / 0 to 300°F
₹1,200.00INR / Each
United Scientific Supplies Red Spirit-Filled Thermometer, 12" Length, Partial Immersion, -20 to 110°C / 0 to 230°F 7890958 Mfr # THPCF1
COMPARE
12 Red spirit Red Spirit-Filled Thermometer, 12" Length, Partial Immersion, -20 to 110°C / 0 to 230°F
₹1,200.00INR / Each
United Scientific Supplies Red Spirit-Filled Thermometer, 12" Length, Partial Immersion, -20 to 150°C / 0 to 300°F 7890959 Mfr # THPCF2
COMPARE
12 Red spirit Red Spirit-Filled Thermometer, 12" Length, Partial Immersion, -20 to 150°C / 0 to 300°F
₹1,200.00INR / Each
United Scientific Supplies Red Spirit-Filled Thermometer, 6" Length, Total Immersion, -20 to 110°C / 0 to 235°F 7890960 Mfr # THPC06
COMPARE
6 Red spirit Red Spirit-Filled Thermometer, 6" Length, Total Immersion, -20 to 110°C / 0 to 235°F
₹930.00INR / Each
Showing 1- 9 of 9