United Scientific Supplies Red Spirit-Filled Thermometer, 12" Length, Total Immersion, -20 to 110°C
Representative image only.
United Scientific Supplies Red Spirit-Filled Thermometer, 12" Length, Total Immersion, -20 to 110°C
| Mfr: THTC01| Item: 7890952
Kerosene-based column is safer than mercury
₹1,100.00INR / Each
₹1,100.00INR / Each
Product Description
Key Features
  • Partial or total immersion to fit your unique application
  • Kersone-based red spirit is safer to use
  • Include no-roll sleeves to prevent accidental breakage
More About this Item
Red spirit-filled thermometers provide increased safety with economic pricing. These thermometers have 6 to 7 mm outer diameters and the bulbs have equal or smaller diameters than the stems. All thermometers include a no-roll sleeve to stop thermometers rolling off of lab benches.
Partial immersion thermometers are useful if you have a shallow container. These thermometers have a small indicator ring that denotes the depth to which the thermometer should be submerged to obtain an accurate temperature reading.
Total immersion thermometers need to be submerged deep enough so the desired temperature is at the meniscus.
Specifications
  • Min Temperature (° C)-20
  • Max Temperature (° C)110
  • Thermometer FillRed spirit
  • Overall Length (InchShort)12
  • Overall Length (mm)304.8
  • DescriptionRed Spirit-Filled Thermometer, 12" Length, Total Immersion, -20 to 110°C