United Electric Electronic Temperature Switches

The Preferred Choice for Plant Instrumentation Upgrades
  • Legendary UE Reliability
  • Hazardous Location
  • Class I, Div 2; Class II, Div. 2; Class III
  • Worldwide approvals and certifications
  • Large process display
  • 100% programmable set-point and deadband
  • Terminal block wiring
  • NEMA 4X / IP66 Die cast aluminum enclosure, epoxy powder coated, & gasketed
421,000.00 - ₹468,000.00INR / Each

2 variations of this product are available.

See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Min Temperature (° F) Description
Availability Pricing
United Electric 2W3A00-T-R1 Temperature Switch, -50/450F; 115V/100 Ma, 6' PTFE 3900751 Mfr # 1XSWHLTR1
COMPARE
450 -50 Temperature Switch, -50/450F; 115V/100 Ma, 6' PTFE
₹421,000.00INR / Each
United Electric 4W3A01-T-H1 Temperature Switch, -50/1000F; 115V/240V/10A, 6' Mi 3900753 Mfr # 1XSWHHTH1
COMPARE
1000 -50 Temperature Switch, -50/1000F; 115V/240V/10A, 6' Mi
₹468,000.00INR / Each
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat