Unico Scanning UV/Visible Spectrophotometers
Advanced spectral analysis for DNA, protein, kinetics, and more!
  • Sophisticated methods with graphic display
  • Programmed GLP verification protocols — use date/time stamping for documentation
  • Transfer data directly to printer using parallel port or interface with computer using RS-232 port
  • Scan speed is adjustable from 75 to 1000 nm/min
Our economical scanning spectrophotometers produce first-rate results using any UV/visible analytical method. High-quality components and systems enable superior resolution, accuracy, and reproducibility. Specify method parametric setup using simple on-board keypad controls or from computer. View spectral graphics on backlit LCD. Sample compartment accommodates up to 8 cuvettes with pathlengths up to 100 mm.
Store up to 30 user methods in non-volatile memory. Programmed methods include DNA/protein ratios (260/280 nm or 260/230 nm). Specify up to 10 wavelengths for multiple sample analysis or use the GLP protocol to verify wavelength and transmittance. Study reaction kinetics for up to 12 hours with sampling intervals down to every 0.5 seconds; use post-run re-scaling and curve/segment targeting to refine rate calculations.
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Bandwidth Max Wavelength (nm) Power (VAC)
Availability Pricing
Unico Scanning Single-Beam UV/Visible Spectrophotometer; 115 VAC 8305730 Mfr # CP-IS28
COMPARE
4.0 nm 1100
Call For Price
Unico Scanning Single-Beam UV/Visible Spectrophotometer; 230 VAC 8305735 Mfr # CP-IS28E
COMPARE
4.0 nm 1100
Call For Price
Unico Scanning Double-Beam UV/Visible Spectrophotometer; 115 VAC 8305780 Mfr # CP-IS48
COMPARE
1.8 nm 1100