Uni-Capsette Cassettes
  • Fit standard stainless steel and disposable plastic base molds
  • Strategically designed slots for efficient solvent flow and drainage
  • Float and remain closed throughout processing
  • Rigid construction eliminates "chattering" during sectioning
  • Acetal copolymer plastic fabrication resists heat, solvents, decalcifying solutions, and effects of microwave processing
  • Accepted by all current automatic cassette labelers
  • Uni-Capsette cassettes have detachable plastic lids to simplify embedding
17,100.00INR / Pkg of 500
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Color
Availability Pricing
Bio Plas Uni-Capsette 6050 Cassettes, White, 500/pack 4863000 Mfr # 6050
COMPARE
White
₹17,100.00INR / Pkg of 500
Bio Plas Uni-Capsette 6053 Cassettes, Green, 500/pack 4863003 Mfr # 6053
COMPARE
Green
₹17,100.00INR / Pkg of 500
Bio Plas Uni-Capsette 6054 Cassettes, Blue, 500/pack 4863004 Mfr # 6054
COMPARE
Blue
₹17,100.00INR / Pkg of 500
Bio Plas Uni-Capsette 6062 Cassettes, Lime, 500/pack 4863009 Mfr # 6062
COMPARE
Lime
₹17,100.00INR / Pkg of 500