Bio Plas Uni-Capsette 6054 Cassettes, Blue, 500/pack
Representative image only.
Bio Plas Uni-Capsette 6054 Cassettes, Blue, 500/pack
Bio Plas | Mfr: 6054| Item: 4863004
₹17,100.00INR / Pkg of 500
₹17,100.00INR / Pkg of 500
Product Description
Key Features
  • Fit standard stainless steel and disposable plastic base molds
  • Strategically designed slots for efficient solvent flow and drainage
  • Float and remain closed throughout processing
  • Rigid construction eliminates "chattering" during sectioning
  • Acetal copolymer plastic fabrication resists heat, solvents, decalcifying solutions, and effects of microwave processing
  • Accepted by all current automatic cassette labelers
  • Uni-Capsette cassettes have detachable plastic lids to simplify embedding
Specifications
  • ColorBlue
  • DescriptionCassettes, Blue, 500/pack