Tygon® 2475 IB High Pressure Thermoplastic Tubing
Ideal for high pressure applications
  • Protects sensitive fluids from oxidation
  • Low surface energy reduces wetting
  • Free from added plasticizers
  • Meets USP Class VI criteria
  • Ideal for general-purpose, laboratory, and single-use applications
  • Free technical application support available to assist with tubing selection
Constructed with reinforced textile and a smooth inner surface, the Tygon® 2474 I.B. tubing is able to withstand higher pressure applications such as single use UF/DF and chromatography systems. The tubing is formulated without the addition of softening agents, reducing potential contamination of sensitive fluids in biopharmaceutical applications.
Due to the absence of plasticizers, the tubing can stand up to aggressive acids, bases, and solvents that would react with or extract plasticizer from a standard PVC tube. The clear tubing allows you to visually inspect the fluid path to ensure consistency of flow. The tubing has a lower gas permeation than standard silicone tubing, allowing you to protect sensitive fluids from oxidation.
In addition, the tubing reduces wetting of aqueous substances allowing for higher recovery of fluids, reduces microbial buildup, and has very low absorption of aqueous substances.
Applications include deionized water systems, aseptic fill lines, cell harvest and media processing, water for injection transport, preservative fill lines, chromatography and UF/DF systems, pharmaceutical and cosmetic processing, and chemical transfer.
Call For Price
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (ft) Tubing ID (in) Tubing OD (in)
Availability Pricing
Tygon 2475 IB Tubing,  1/4" ID x 1/2" OD; 50 Ft 5011382 Mfr # ACX00019
COMPARE
50 1/4 1/2
Call For Price
Tygon 2475 IB Tubing,  3/8" ID x 5/8" OD; 50 Ft 5011383 Mfr # ACX00029
COMPARE
50 3/8 5/8
Call For Price
Tygon 2475 IB Tubing,  1/2" ID x 3/4" OD; 50 Ft 5011384 Mfr # ACX00038
COMPARE
50 1/2 3/4
Call For Price
Tygon 2475 IB Tubing,  5/8" ID x 7/8" OD; 50 Ft 5011385 Mfr # ACX00046
COMPARE
50 5/8 7/8
Call For Price
Tygon 2475 IB Tubing,  3/4" ID x 1-1/16" OD; 50 Ft 5011386 Mfr # ACX00054
COMPARE
50 3/4 1 1/16
Call For Price
Tygon 2475 IB Tubing,  1" ID x 1-3/8" OD; 25 Ft 5011387 Mfr # ACX42064
COMPARE
25 1 1 3/8
Call For Price
Tygon 2475 IB Tubing,  1/4" ID x 1/2" OD; 10 Ft 5011388 Mfr # ACX06074
COMPARE
10 1/4 1/2
Call For Price
Showing 1- 7 of 7