Troemner Mass Accessories - Analytical Weight Forks
Various materials to suit your applications
We offers a variety of forks of various materials to suit your applications.
Plastic weight forks are constructed of high quality polycarbonate and are designed specific to the size and style of the weight.
Wooden weight forks are ideal for 200 - 500 g analytical precision weights.
5,800.00 - ₹7,500.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Troemner WA001 Wood Weight Fork for 200g - 500g or 1/2 lb to 1 lb Analytical Precision Weights 0102796 Mfr # WA001
COMPARE
Wood Weight fork for 200g - 500g or 1/2 Lb to 1 Lb Weights
₹7,500.00INR / Each
Troemner WA219 Polycarbonate Analytical Weight Fork for 500g - 2kg Analytical Precision Weights 0102797 Mfr # WA219
COMPARE
Polycarbonate Analytical Weight Fork for 500g - 2kg Analytical Precision Weights
₹5,800.00INR / Each