Troemner WA219 Polycarbonate Analytical Weight Fork for 500g - 2kg Analytical Precision Weights
Representative image only.
Troemner WA219 Polycarbonate Analytical Weight Fork for 500g - 2kg Analytical Precision Weights
Troemner | Mfr: WA219| Item: 0102797
Various materials to suit your applications
₹5,800.00INR / Each
₹5,800.00INR / Each
Product Description
More About this Item
We offers a variety of forks of various materials to suit your applications.
Plastic weight forks are constructed of high quality polycarbonate and are designed specific to the size and style of the weight.
Wooden weight forks are ideal for 200 - 500 g analytical precision weights.
Specifications
  • Mass CertificationNot applicable
  • DescriptionPolycarbonate Analytical Weight Fork for 500g - 2kg Analytical Precision Weights